missing translation for 'onlineSavingsMsg'
Learn More

Fisher Healthcare™ SHUR/Clean™ Active Sloped-Front Fume Hoods for Grossing

Catalog No. p-4488673
Change view
Click to view available options
Voltage:
100/115 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
22047201 100/115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22047201 Supplier Fisher Healthcare™ Supplier No. 22047201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Bench-top fume hoods to help manage routine manual histological processes

The Sloped-Front Fume Hood features two 174 cfm high-speed fans that pull contaminated air through two 8 x 8 in. prefilters to remove dust and particulate matter and two 8 x 8 in. charcoal filters to provide the protection needed for hazardous fume absorption. Double-hinged front sash provides convenient access for multiple applications.

  • Microprocessor controlled
  • Double-hinged sash
  • Two prefilters for particulate removal and two carbon filters for fume containment
  • Two constant-speed 174 cfm blowers
  • Face velocity: 90 fpm with sash completely down
  • 22 in. fluorescent 13W T-5 bulb
  • External dimensions: 36W x 20D x 28 in.H (91 x 51 x 71cm)
  • Internal dimensions: 34W x 19D x 19 in.H (86 x 48 x 48cm)
  • Weight: 67 lb. (31kg)
  • Model SC-36S-F (for Grossing): contains two carbon filters (SC-36S-FF) for formaldehyde fume containment

Specifications

Product Type Active Sloped-Front Ductless Fume Hood
Depth (English) 20 in.
Depth (English) Interior 19 in.
Depth (Metric) 51 cm
Depth (Metric) Interior 48 cm
Dimensions (D x W x H) 19 x 34 x 19 in. (48 x 86 x 48 cm)
For Use With (Application) Grossing
Height (English) 28 in.
Height (English) Interior 19 in.
Height (Metric) 71 cm
Height (Metric) Interior 48 cm
Voltage 100/115 V
Weight (English) 67 lb.
Weight (Metric) 31 kg
Width (English) 36 in.
Width (English) Interior 34 in.
Width (Metric) 91 cm
Width (Metric) Interior 86 cm
Show More Show Less