missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Ultimate N-DEX™ 9905 Nitrile Gloves

Catalog No. 296351166
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296351166 Large
296351164 Small
296351165 Medium
296351167 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296351166 Supplier SHOWA™ Supplier No. 9905PFL
Only null left

Ultimate N-DEX™ Nitrile Gloves are engineered with extended-length and a thicker mil for tactile precision, maximum comfort and protection in a wide-range of applications.

  • Thicker mil provides added barrier protection
  • 11 in. longer cuff to protect wrists
  • Second skin fit and feel
  • Rolled cuff prevents dirt from entering the glove
  • Recommended for agricultural, food services, municipal services, pharmaceutical and utilities
  • Made in the USA

Specifications

Certifications/Compliance NFPA 1999, 21 CFR 170-199, ASTM
Applications Industrial
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
Model 9905PFL
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less