missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 377 Nitrile Foam Coated Chemical Resistant Gloves

Catalog No. 19172209
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19172209 Large
19172210 X-Large
19020701 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19172209 Supplier SHOWA™ Supplier No. 377L08
Only null left

Gloves feature a nitrile foam coating for enhanced grip capabilities in slippery environments, with a seamless liner and elasticated cuff to keep hands warm and protected.

  • Chemical resistant gloves with ultra-tactile grip and durability
  • Nitrile foam coating enhances grip capabilities in slippery environments
  • Seamless liner and elasticated cuff to keep hands warm and protected

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Nitrile Foam
Color Blue, Black
Hand Specific
Material Nylon, Polyester
Product Type Dip-coated Gloves
Size Large
EN388 Standard 4121X
For Use With (Application) Agriculture, Construction, and Manufacturing
Glove Cuff Color Gray
Glove Cuff Style Elasticated
Series 377
Show More Show Less