missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Atlas™ 300 Textured Latex Coated Seamless Knit Work Gloves

Catalog No. p-3849190
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19014300 Small
19014301 Medium
19014302 Large
19014303 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19014300 Supplier SHOWA™ Supplier No. 300S07
Only null left

Gloves are ergonomically designed with a lightweight coating to ensure maximum dexterity, while also enhancing grip capabilities.

  • Ergonomic design ensures a secure fit and long-lasting comfort
  • Lightweight coating enhances grip capabilities
  • Elastic knit wrist cuff helps prevent dirt and debris from entering the glove
  • Open-back ensures breathability and reduces perspiration

Specifications

Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Blue, Gray
Exterior Finish Fully Textured
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 2142X
For Use With (Application) Agriculture, Construction, Municipal Services, and Warehousing
Glove Cuff Style Knit
Series ATLAS™ 300
Show More Show Less