missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 490 Triple-Dipped PVC-Coated Cold Resistant Gloves

Catalog No. 19270241
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19270241 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19270241 Supplier SHOWA™ Supplier No. 490M08

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves feature a triple-dipped PVC coating for protection from oil, grease, and chemicals that will remain flexible at temperatures as low as -20°C.

  • Gloves provide protection and maintain flexibility at temperatures as low as -20°C
  • Triple-dipped PVC coating with a rough texture ensures tactile precision
  • Extended forearm protection from liquids such as oil, grease, and chemicals
  • Ergonomic, form-fitting shape reduces hand fatigue over long periods of wear

Specifications

Coated Area Full
Coating Material PVC
Color Blue
Exterior Finish Fully Textured
Hand Specific
Size Medium
For Use With (Application) Chemical Spray/Treatment, Washing, and Cleaning
Glove Cuff Style Safety
Liner Material Acrylic
Series 490
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less