Research Products Corporation Shorty Serological Pipet, Sterile, Individually Wrapped, 5ml, Sterile, 200 per Case
Catalog No. 50998505
Mini-Serological pipettes are only nine inches long. This shorter length provides a greater convenience when pipetting in tissue culture hoods and tight spaces. Pipettes made from transparent medical grade polystyrene and feature easy to read graduation, lint free plugs and color coded rings for quick identification. Pipettes are individually wrapped and sterile.
Catalog No.
50998505
Supplier
Research Products CorporationSupplier No.
RPI 218400
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mini-Serological pipettes are only nine inches long. This shorter length provides a greater convenience when pipetting in tissue culture hoods and tight spaces. Pipettes made from transparent medical grade polystyrene and feature easy to read graduation, lint free plugs and color coded rings for quick identification. Pipettes are individually wrapped and sterile.