Inmark STP-302 CATEGORY B AMBIENT INSULATED SHIPPING SYSTEM (UN 3373)
Catalog No. NC0090492
STP-302 Category B / Exempt Shipping System, Insulated, Outer Box OD 8.25 x 7 x 6.25in. pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings, Insulated Chest ID 6 x 4.5 x 4in., 8/Case
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
STP-302 Category B / Exempt Shipping System, Insulated, Outer Box OD 8.25 x 7 x 6.25in. pre-printed with Biological Substance, Category B, UN 3373 and Exempt Human/Animal Specimen Markings, Insulated Chest ID 6 x 4.5 x 4in., 8/Case