Learn More
BINDER™ Shipping Pallet
Catalog No. 502875675
Change view
Click to view available options
For Use With (Equipment):
KB260, KB470, KB720, KB1060
KBF260, KBF470, KBF720, KBF1060
KBFPRO260, KBFPRO470, KBFPRO720
KBPRO260, KBPRO470, KBPRO720
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
502875675 KBPRO260, KBPRO470, KBPRO720
502875742 KB260, KB470, KB720, KB1060
502875613 KBF260, KBF470, KBF720, KBF1060
502875625 KBFPRO260, KBFPRO470, KBFPRO720
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 502875675 Supplier BINDER™ Supplier No. 80122546
Only null left
Shipping pallet with unloading ramp for installing unit without forklift
For
Shipping pallet with unloading ramp for easy unloading of chamber from shipping pallet.
Specifications
For Use With (Equipment) KBPRO260, KBPRO470, KBPRO720
Type Platform
Material Fiber