Learn More
BINDER™ Shipping Pallet
Description
For
Shipping pallet with unloading ramp for easy unloading of chamber from shipping pallet.
Specifications
Specifications
| For Use With (Equipment) | KBF260, KBF470, KBF720, KBF1060 |
| Type | Platform |
| Material | Fiber |