missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Shelves for FreeZone™ Freeze Dryers

Catalog No. 16108571
Change view
Click to view available options
Size:
Short
Tall
No. of Shelves:
3
5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size No. of Shelves
16108571 Tall 5
16108570 Short 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16108571 Supplier Labconco™ Supplier No. 7317200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.

  • Removable, 10 in. diameter aluminum shelves with 1/4 in. rim
  • Shelves slide into stainless steel support
  • Includes 3 rubber feet
  • Design allows for flexible shelf height spacing
  • Shelf can be placed in freezer to prefreeze sample for quick loading
  • Designed to fit inside Drying Chambers (sold separately)
  • Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control on models manufactured 2016 and earlier and up to +60°C (+140°F) on models 2017 and later (contact your Fisher Customer Service Representative for details).

Specifications

Type Product Shelves
Capacity (Metric) 250 L
No. of Shelves 5
Material Stainless Steel
Size Tall
For Use With (Application) Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications
For Use With (Equipment) FreeZone™ Drying Chambers
Includes Five aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet
Width (English) 10.4 in.
Height (English) 17.5 in.
Height (Metric) 44.45 cm
Length (English) 10.9 in.
Length (Metric) 27.7 cm
Shipping Weight (English) 7 lb.
Weight (English) 5.5 lb.
Show More Show Less