missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Shelves for FreeZone™ Freeze Dryers
Description
Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications.
- Removable, 10 in. diameter aluminum shelves with 1/4 in. rim
- Shelves slide into stainless steel support
- Includes 3 rubber feet
- Design allows for flexible shelf height spacing
- Shelf can be placed in freezer to prefreeze sample for quick loading
- Designed to fit inside Drying Chambers (sold separately)
- Also available: Heated shelves that include ambient to +40°C (+104°F) variable heat control on models manufactured 2016 and earlier and up to +60°C (+140°F) on models 2017 and later (contact your Fisher Customer Service Representative for details).
Specifications
Specifications
| Type | Product Shelves |
| Capacity (Metric) | 150 L |
| No. of Shelves | 3 |
| Material | Stainless Steel |
| Size | Short |
| For Use With (Application) | Accommodate bulk samples, microtiter plates, vials, or serum bottles for bulk drying applications |
| For Use With (Equipment) | FreeZone™ Drying Chambers |
| Includes | Three aluminum shelves with 1/4 in. rims, one stainless steel support, and three rubber feet |
| Width (English) | 10.4 in. |
| Height (English) | 9.8 in. |
| Show More |