Learn More
Thermo Scientific™ Stainless Steel Shelf Kit 
Catalog No. 15467330
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
15467330 Stainless Steel
1 options
Catalog No. 15467330 Supplier Thermo Scientific™ Supplier No. 190884
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Stainless Steel shelf kit includes 1 Stainless Steel shelf with support channels for installation into Forma™ and Steri-Cycle™ CO2 incubators
Get maximum flexibility for your incubator or environmental chamber with these Thermo Scientific™ Incubator and Environmental Chamber accessories.
Specifications
Dimensions (D x W) Shelf 18.4 x 18.4 in. (46.8 x 46.8 cm)
For Use With (Equipment) Steri-Cycle™ Direct Heat, Steri-Cycle™ Water Jacket Series 2, Steri-Cycle™ Water Jacket Series 3, and Steri-Cycle™ 185 Pro Series CO2 Incubators
Lighting LED
Material Stainless Steel
No. of Shelves 1
Type Shelf Kit