Sheep Adrenocorticotropic Hormone (Acth) Elisa Kit for detecting Adrencocorticotropic Hormone (Acth) in serum, plasma. Species Reactivity: Sheep (Ovis aries). Detection Range: 0.097-400pg/ml. Sensitivity: 0.097pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.