missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Sharps Container, Quart (1L)
Description
- Clearly labeled with the Biohazard warning on one side.
- Provides safe disposal of syringes, needles and other small sharps.
- Stackable to reduce storage space before use.
- 1 L Capacity quart with a clear lid to view fill level.
- Slide access lid provides temporary and final closures for safety and security.
- Built-in needle removal port, prevents direct contact with sharps.
- Compatible with the Droplet blood collection tray, Totemax™ and blood collection tray.
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Color | Red |
| Dimensions (L x W x H) | 4.5 x 4.5 x 7.5 in. |
| Disposable | Reusable |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Material | Polypropylene |
| Product Type | Sharps Disposal Container |