missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sharps Container, Quart (1L)

Catalog No. 14127525
Change view
Click to view available options
Color:
Red
Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14127525 Yellow
14127537 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14127525 Supplier Fisherbrand™ Supplier No. 120178F2
Only null left

Easy to use and transport. Can be used alone or with Fisherbrand Blod Collection Trays.

  • Clearly labeled with the Biohazard warning on one side.
  • Provides safe disposal of syringes, needles and other small sharps.
  • Stackable to reduce storage space before use.
  • 1 L Capacity quart with a clear lid to view fill level.
  • Slide access lid provides temporary and final closures for safety and security.
  • Built-in needle removal port, prevents direct contact with sharps.
  • Compatible with the Droplet blood collection tray, Totemax™ and blood collection tray.

Specifications

Capacity (Metric) 1 L
Color Yellow
Dimensions (L x W x H) 4.5 x 4.5 x 7.5 in.
Disposable Reusable
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Polypropylene
Product Type Sharps Disposal Container