missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sharps Container

The Fisherbrand Sharps Container, designed for higher volume usage in laboratories, doctors' offices, and clinics for larger biohazardous waste items, enables laboratory or medical professionals to temporarily seal the container for secure internal transportation.

Supplier:  Fisherbrand™ 0320150FFS

 View more versions of this product

Catalog No. 22-730-467

Only null left
Explore available promotions

Description

Description

  • Provides safe disposal of needles and syringes.
  • The container is available in various sizes to accommodate different requirements, and it features different lid designs to suit various preferences.
Specifications

Specifications

ISO 23907, OSHA 29 CFR 1910.1030, FDA 510k
Flip Top Lid
2 gal.
10.3 x 7 x 9.7 in.
Yes
20/Cs
Yes
Not Autoclavable
10.3 in.
Case
Sharps Disposal Container
9.7 in.
Clear, White
Red
Single-use
8.2 x 2
Rectangle
Non-sterile
7 in.
9.7 in.
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions