Learn More
Fine Science Tools SHARP/BLUNT SCISSORS DFS Item
Supplier:  Fine Science Tools 9140112
Tip Shape: Straight/ Tips: Sharp-Blunt/ Cutting Edge: 40mm/ Alloy / Material: Student Stainless Steel/ Length: 12cm/ Serrated: No/ Feature: Student Quality
Catalog No. NC9573906
Only null left
Explore available promotions