Learn More
Epredia™ Xylene Substitute
Catalog No. 6764506
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6764506 4 x 1 gal.
9990505 1 gal.
2860075 5 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 6764506 Supplier Epredia™ Supplier No. 6764506
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.
  • Not sensitizing to normal skin
  • Evaporation rate similar to xylene
  • Virtually odorless aliphatic hydrocarbon—no citrus aroma
  • Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications
Product Type Xylene Substitute
Quantity 4 x 1 gal.