Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Catalog No. 2861813
Change view
Click to view available options
Quantity:
59 mL
5 x 59 mL
Color:
Black
Blue
Green
Multiple
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
2861813 5 x 59 mL Multiple
2860350 59 mL Blue
2860356 59 mL Black
2960357 59 mL Yellow
2860358 59 mL Red
2860362 59 mL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2861813 Supplier Epredia™ Supplier No. 3120123
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Color Multiple
Product Type Tissue Marking Dye
Quantity 5 x 59 mL
Includes 1 bottle each of blue, black, yellow, red, green; 50 applicator sticks; storage base