missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit

Catalog No. 2860358
Change view
Click to view available options
Quantity:
59 mL
5 x 59 mL
Color:
Black
Blue
Green
Multiple
Red
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
2860358 59 mL Red
2861813 5 x 59 mL Multiple
2860350 59 mL Blue
2860356 59 mL Black
2960357 59 mL Yellow
2860362 59 mL Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2860358 Supplier Epredia™ Supplier No. 3120127
Only null left

Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.

  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning

Ordering Information:

Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).

Specifications

Color Red
Product Type Tissue Marking Dye
Quantity 59 mL
For Use With (Application) Healthcare