Learn More
Epredia™ TBD-1™ Decalcifier
Catalog No. 2860079
Change view
Click to view available options
Quantity:
2 Pk.
Catalog No. Quantity
2860079 2 Pk.
1 options
Catalog No. 2860079 Supplier Epredia™ Supplier No. 6764001
Process bone marrow biopsies in as little as 15 to 30 minutes with this rapid hydrochloric acid decalcifier.
  • Process thin pieces of dense bone in 3 to 12 hours
  • Permits utilization of routine end point tests, such as calcium oxalate
  • Protects cells from actions of acids with added ingredients
Specifications
Product Type Decalcifying Solution
Quantity 2 Pk.