missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Straight Blunt/Sharp Dissecting Scissors, premium, 6.5 in. (16.5cm)
Epredia™ Shandon Straight Blunt/Sharp Dissecting Scissors are available in Premium and Standard grades.
Supplier: Epredia™ 728252
Description
- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards, producing the finest instruments available
- Guaranteed to be free from defects in material and workmanship
Chemical Identifiers
| Dissecting Scissors | |
| Straight | |
| Blunt/Sharp |
Specifications
| Dissecting Scissors | |
| Premium | |
| 14 cm |
| Straight | |
| 5.5 in. | |
| Blunt/Sharp |