Learn More
Epredia™ Rapid-Fixx™ Formalin Fixative
Catalog No. 2860044
Change view
Click to view available options
Quantity:
2 x 1 L
Catalog No. Quantity
2860044 2 x 1 L
1 options
Catalog No. 2860044 Supplier Epredia™ Supplier No. 6764212
Only null left
Obtain quick results with this formalin. Excellent for antigenic preservation, avoids the need for enzyme digestion, and rapidly penetrates into tissue.
Specifications
Quantity 2 x 1 L