Learn More
Epredia™ Shandon™ Rapid-Chrome Papanicolaou Staining Kit
Description
Includes: five-slide holder, portable tray, rinse water, Gill 2 Hematoxylin, bluing reagent, EA-50, 100% alcohol, OG-6, 95% alcohol, Xylene, mounting medium
Specifications
Specifications
| Type | Papanicolaou Stain |
| Includes | Five-slide holder |