Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Catalog No. 3810409
Change view
Click to view available options
Dimensions (L x W x H):
22x22 to 12x12 mm, 20 mm deep
22 x 22 x 20 mm
22 x 30 x 20 mm
22 x 40 x 20 mm
Shape:
Rectangular
Square
Truncated, tapered
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Shape
3810409 22 x 30 x 20 mm Rectangular
1841 22 x 40 x 20 mm Rectangular
2219 22 x 22 x 20 mm Square
1220 22x22 to 12x12 mm, 20 mm deep Truncated, tapered
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3810409 Supplier Epredia™ Supplier No. 1830
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
  • Constructed of rigid transparent plastic.
Specifications
Product Type Embedding Mold
Material Plastic
Dimensions (L x W x H) 22 x 30 x 20 mm
Disposable Single-use
Shape Rectangular