Learn More
Epredia™ Shandon™ Nu-Clear™ Reagent
Catalog No. 6769008
Change view
Click to view available options
Product Type:
Tissue Processing Reagent (Nu-Clear I)
Tissue Processing Reagent (Nu-Clear II)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
6769008 Tissue Processing Reagent (Nu-Clear I)
6769009 Tissue Processing Reagent (Nu-Clear II)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6769008 Supplier Epredia™ Supplier No. 6769008
Achieve improved cell clarity by removing excess hematoxylin from the slide with Epredia™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.
Key Features 
  • Removes residual hematoxylin staining the glass.
  • Designed to be used on the full line of Epredia Shandon automatic stainers as well as manual methods.
Nu-Clear I
  • Fast-acting solution
  • Recommended to be used as a 10-second step in most protocols
  • Particularly beneficial for use with Harris hematoxylin
  • May be used on the Epredia Varistain™ 24-4 or Shandon Varistain Gemini slide stainers
Nu-Clear II
  • Less concentrated
  • Recommended as a 20-second step
  • May be used on the Epredia Shandon Linistat and other linear stainers
  • Rcommended for use with Gill hematoxylins
Specifications
Product Type Tissue Processing Reagent (Nu-Clear I)