missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon-Mount™
Perform quick, high-quality conventional coverslipping with Shandon-Mount™ permanent mounting medium.
Supplier: Epredia™ 1900333
Description
- Overnight-drying liquid cover slip
- Toluene base
- Refractive index: 1.495 ±0.005 (dry)
- Viscosity: Low (72-120 cps at 22°C)
Specifications
| Mounting Medium | |
| 120 mL |