Learn More
Epredia™ Shandon™ Kwik-Diff™ Stains
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.
Supplier:  Epredia™ 9990705
 View more versions of this product
Catalog No. 28-613-2
Only null left
Description
Description
  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Includes:
Reagent 1: Fixative
Specifications
Specifications
4 L
Fixative
4 L Kwik-Diff Reagent 1, Fixative
Product Suggestions
Product Suggestions