Learn More
Epredia™ Shandon™ Kwik-Diff™ Stains
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.
Supplier:  Epredia™ 9990701
 View more versions of this product
Catalog No. 28-613-1
Only null left
Description
Description
  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Includes:
Reagent 1: Fixative; Reagent 2: Eosin; Reagent 3: Methylene Blue
Specifications
Specifications
125 mL
Kwik-Diff Stains
125 mL Each: Fixative, Eosin, Methylene Blue
Product Suggestions
Product Suggestions