missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Instant Eosin
Description
- Stable and precipitate-free; no filtering
- pH-adjusted for maximum stain uptake
- Economical powder stores indefinitely
- Compatible with all alum Hematoxylin stains
Specifications
Specifications
| Compatibility | Alum Hematoxylin Stains |
| Product Type | Eosin |
| Solution Type | Eosin |
| Form | Liquid |