missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Small Charcoal Filter

Catalog No. 9990610
Change view
Click to view available options
Size: 11.25 x 5.75 x 1.31 in. (29 x 15 x 3.3 cm); Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
9990610 Size: 11.25 x 5.75 x 1.31 in. (29 x 15 x 3.3 cm); Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9990610 Supplier Epredia™ Supplier No. 9990610

For use with Epredia™ Hyperclean Pathology Workstations.

Accessories:
  • Charcoal Filter
  • Fine particle Filters
  • Formaldehyde Neutralizing (Potassium Permanganate) Filter

Specifications

Product Type Charcoal Filter
Description Size: 29 cm x 15 cm x 3.3 cm
For Use With (Equipment) Epredia Hyperclean Pathology Workstations, Models 1, 20, 22, 36, 60
Product Line Hyperclean
Quantity 6 Pk.