missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Grooved Needle Director, 5 in. (12.7cm)

Catalog No. 1100
Change view
Click to view available options
Length (English):
5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
1100 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1100 Supplier Epredia™ Supplier No. 1100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose 5 in. (12.7cm) Epredia™ Shandon™ Grooved Needle Director for quality and precision.

Features:

  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Patterns, dimensions and finish held to exacting standards
  • Guaranteed to be free from defects in material and workmanship

Specifications

Length (English) 5 in.