Learn More
Epredia™ Glyo-Fixx™
Description
- Produces distinct and crisp cellular morphology
- Fixes most antigens without impairing immunoreactivity
- Allows special stain dyes, including silver procedures, to be sharp without background staining
- Presents fine detail in specimen structure
Specifications
Specifications
| Product Type | Fixative (Concentrate) |
| Quantity | 1 gal. (makes 5 gal.) |