missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin

Catalog No. 2860347
Change view
Click to view available options
Quantity:
120 mL container 60 mL filled
3.8 L
5 gal. (19L)
60 mL container 30 mL filled
90 mL
Reconstituted to 5 Liters (Ea.)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2860347 90 mL
6764240 5 gal. (19L)
2860067 3.8 L
2860064 Reconstituted to 5 Liters (Ea.)
2860340 60 mL container 30 mL filled
2860348 120 mL container 60 mL filled
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2860347 Supplier Epredia™ Supplier No. 9990916
Only null left

Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available

Specifications

Capacity 90 mL Specimen Container filled to 45 mL
Packaging Type 125/Cs.
Product Type Formalin
Quantity 90 mL