missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin
Description
- Phosphate-free
- Use for storage of all tissue types
- Stores specimens without excessive shrinking or hardening
- Available premixed, ready-to-use, or in economical concentrate
- Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
- Pre-filled, leak-resistant cups are available
Specifications
Specifications
| Capacity | Reconstituted to 5 gallon (19 L) |
| Packaging Type | Ea. |
| Product Type | Formalin |
| Quantity | 3.8 L |