Learn More
Epredia™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders, Low profile
Eliminate the need to switch to a different knife holder for larger specimens by using the Epredia™ Shandon™ Finesse™ ME/E Series Disposable Blade Holders.
Supplier:  Epredia™ A77510272
Catalog No. 28618310
Only null left
Description
Description
  • Improved clamping mechanism design significantly reduces need for knife holder maintenance and improves reliability
  • Consistent reproducible sections provided
  • Larger clamp plate allows easier section retrieval
  • Clamped blade is fully supported along its entire length
  • Entire top stage moves laterally for safer operation
  • Minimized wear on clamping mechanism
Product Suggestions
Product Suggestions