missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ EZ-Mount™ Mountant

Catalog No. 9999120
Change view
Click to view available options
Capacity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
9999120 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9999120 Supplier Epredia™ Supplier No. SH9999120
Only null left

Inhibit stain fading with Epredia™ EZ-Mount™ Mountant, a clear, xylene-based mounting medium compatible with membrane filters.

  • Refractive index: 1.495 ± 0.005 (dry)
  • Viscosity: High (1280-2790 cps at 22°C)

Specifications

Capacity 500 mL