missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Dual Blade Manual Bone Saw: Replacement Blades

Catalog No. 2861586
Change view
Click to view available options
Description:
Replacement Blades, Coarse
Replacement Blades, Fine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
2861586 Replacement Blades, Coarse
2861587 Replacement Blades, Fine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2861586 Supplier Epredia™ Supplier No. 1001798

Replacement Blades for use with Shandon Dual Blade Manual Bone Saw.

High-quality blades for use with Shandon Dual Blade Manual Bone Saw

Specifications

Description Replacement Blades, Coarse
For Use With (Equipment) Dual Blade Manual Bone Saw (1001797)
Includes Replacement blade set (includes links)
Length (English) 7 in.
Length (Metric) 17.8 cm
Width (English) 9 in.
Width (Metric) 22.9 cm