Learn More
Epredia™ Shandon™ CytoRich™ Red Collection Fluid
Catalog No. p-4530251
Change view
Click to view available options
Quantity:
20 mL
120 mL
500 mL
1 gal.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28618248 20 mL
B9990803 120 mL
B9990801 500 mL
B9990800 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 28618248 Supplier Epredia™ Supplier No. B9990802
Only null left
Allow for more accurate evaluation of cytological features with Epredia™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry
Specifications
Product Type Collection Fluid
Quantity 20 mL