Learn More
Epredia™ Cryochrome™ Embedding Resin
Catalog No. 2861147
Change view
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Sampler Colors
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
2861147 Cryochrome Green
2861145 Cryochrome Blue
2861095 Cryochrome Sampler Colors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2861147 Supplier Epredia™ Supplier No. 9990428
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Product Type Cryochrome Green
Quantity 120mL