Learn More
Epredia™ Cryochrome™ Embedding Resin
Catalog No. 2861095
Change view
Click to view available options
Product Type:
Cryochrome Blue
Cryochrome Green
Cryochrome Sampler Colors
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
2861095 Cryochrome Sampler Colors
2861145 Cryochrome Blue
2861147 Cryochrome Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 2861095 Supplier Epredia™ Supplier No. 9990426
Only null left
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Product Type Cryochrome Sampler Colors
Quantity 120mL