missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ ClearVue™ Coverslipper

Boost speed and accuracy in busy laboratories by automating the coverslipping workload with the Epredia™ ClearVue™ Coverslipper.

Supplier:  Epredia™ A79200001

Catalog No. 28618381

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Simply place a basket of slides onto the load rail and close the door — the ClearVue Coverslipper does all the rest.

Compatibility

  • Offers compatibility with a variety of stainers to accommodate just about any lab setting
  • Accepts slides directly from the Epredia™ Varistain™ Gemini ES and Varistain™ 24-4, Sakura™ DRS2000 and Leica™ Auto-Stainer
  • Permits loading of slide baskets from any of these stainers in a mix-or-match fashion without requiring operator adjustments to the coverslipper

Specimen Recognition

  • Automatically recognizes different types of slide preparations and delivers the correct amount of mountant for coverslipping each
  • Handles both histology and cytology samples simultaneously without user interaction while still delivering right amount of mountant

Load-on-Demand

  • Handles up to 11 slide baskets at the same time with load-on-demand capabilities
  • Accepts baskets loaded one at a time or up to five baskets loaded together

Optical Recognition

  • Possesses the unique ability to optically recognize and position slides during the coverslipping process
  • Recognizes the position of each slide in a basket, removes each slide with a set of slide-grippers and returns every slide back, while maintaining slide positioning

User Interface

  • Intuitive user interface features a touchscreen panel
  • Software monitors the type of slide preparation, size and number of slips dispensed and the number of baskets waiting to be coverslipped

Safety Features

  • System is not operational when any of its doors are open, preventing mechanical mishaps
  • Downdraft ventilation protects the user when loading and unloading slide baskets
  • Charcoal filter removes any vapors before they can escape the unit
  • Removable debris and purge trays make cleanup a breeze

Physical Specifications

  • Dimensions (D x W x H): 57.5 x 64.5 x 50 cm (22.75 x 25.5 x 19.75 in.)
  • Weight: 110 lb.
  • Power Requirements: 100-240 VAC, 50/60 Hz

Includes:

  • User guide (English, part number A79210100)
  • Case of two ClearVue coverslip hoppers
  • 24 x 50 mm, No. 1.5–1000 slips (A79210050)
  • Set of three ClearVue slide baskets (Gemini-compatible) with black sliders (A79210064)
  • Set of two mountant purge trays (A79210079)
  • Ventilation filter (9990610)
  • Set of two soft hair brushes (P12257)
  • Set of three suction cups (A79210081)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions