Learn More
Epredia™ Shandon™ Cell-Fixx™ Spray Fixative
Includes Carbowax™ solution, which coats the smear to maintain cellular detail Prevents air drying artifact and cell loss.
Supplier:  Epredia™ 6768326
Catalog No. 2860041
Only null left
Description
Description
Specifications
Specifications
Fixative Spray
6 x 50 mL
Product Suggestions
Product Suggestions