Learn More
Epredia™ Shandon™ Bluing Reagent
Catalog No. 2860081
Change view
Click to view available options
Quantity:
4L
2 x 1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2860081 2 x 1L
6769002 4L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2860081 Supplier Epredia™ Supplier No. 6769001
Only null left
Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.
  • Ready-to-use
  • Thoroughly pH-controlled for completely effective bluing
Specifications
Type Bluing Reagent
Quantity 2 x 1L