Learn More
Shake-N-Cast™ Impression Kit
Catalog No. S99541
Change view
Click to view available options
Impression kit
Catalog No.
S99541 Impression kit
1 options
Catalog No. S99541 Supplier Customer Channels Group (Ccg) Supplier No. SNC42
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
2 lb. (0.9kg) dental stone and 14 oz. (414mL) liquefier, factory-sealed in a heavy-duty plastic pouch
Self-contained and pre-measured to eliminate all the mess and guesswork.
  • Mixture is always the right consistency
  • Simply apply pressure to break the water capsule, shake and knead the pouch for up to two minutes, cut off a corner and pour into impression
Order Info
One kit is sufficient for casting a full-sized adult shoe print.
Specifications
For Use With (Application) Casting a full-sized, adult shoe print
Includes 2 lb. dental stone and 14 oz. liquefier, factory-sealed in a heavy-duty plastic pouch
Weight (English) 42 oz.
Weight (Metric) 1190.6 g
Product Type Shake-N-Cast™ Impression Kit