Learn More
World Precision SGE Syringes with Replaceable Beveled Needles-SGE010RNS
Catalog No. NC1463899
Encompass
A 20 bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.
Catalog No. NC1463899 Supplier World Precision Supplier No. SGE010RNS
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A 20 bevel tip is the standard general purpose needle tip style supplied with SGE syringes. The style is designed for optimum septum penetration and prevention of septum coring.