missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ eVol™ XCHANGE Syringes

XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.

$472.32 - $472.32

Specifications

For Use With (Application) Liquid handling procedure
Length (English) 1.9 in.
Length (Metric) 50 mm
Product Type eVol™ Syringe
Length (Metric) Needle 50 mm
View More Specs

Products 1
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Quantity Price Quantity  
03-350-320
View Documents
Trajan™
2910029
100 μL 1 Pk.
Each for $472.32
Only null left
 
Description

Description

  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available
Specifications

Specifications

Liquid handling procedure
50 mm
50 mm
1.9 in.
eVol™ Syringe
Bevel
Videos
SDS
Documents

Documents