missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SFM4Transfx-293 Media
Description
- ADCF
- Supports high transfection efficiency
- Metabolically designed for high cell yield and recombinant protein/vector production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
Specifications
| Classification | None |
| Form | Liquid |
| Product Type | SFM4Transfx-293 |
| Sterility | Sterile |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | Poloxamer 188, Sodium Bicarbonate |
| Without Additives | No Protein, No Phenol Red, No HEPES, No Glutamine |
| Quantity | 1000 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2° to 8°C, away from light |