Learn More
Expression Systems, Llc Sf9 cells, Frozen in ESF 921, 50M cells/vial DFS Item
Supplier:  Expression Systems Llc 94001F
Expression Systems offers insect cell lines adapted to serum-free suspension culture in either ESF 921 or ESF AF. The availability of pre-adapted cells saves significant time and expense associated with the adaptation of cultures.
The Sf9 insect cell line is a clonal isolate derived from the parental Spodoptera frugiperda cell line IPLB-Sf-21-AE. Sf9 cells are adapted for suspension culture in ESF 921 or ESF AF and are available as a frozen vial or suspension culture. Sf9 cells are commonly used to produce recombinant baculoviral stocks and to produce recombinant proteins.
Recombinant protein expression from a variety of expression platforms
Good growth in adherent or suspension culture
Small, regular size that generates even monolayers and plaques
Quality and performance testing
To download Instructions for Use please visit: www.expressionsystems.com
Catalog No. NC0633748
Only null left
Explore available promotions