missing translation for 'onlineSavingsMsg'
Learn More

Seward™ Stomacher™ Bag Racks

Catalog No. 142853
Change view
Click to view available options
Holds:
Bags
For Use With (Equipment):
Stomacher 3500
Stomacher 400
Stomacher 400 lab blenders
Stomacher 80Biomaster
Dimensions (L x W x H):
15 cm x 42 cm x 35 cm
35 cm x 10 cm x 22 cm
8 cm x 10.5 cm x 19 cm
30 x 11 x 12 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds For Use With (Equipment) Dimensions (L x W x H)
142853 Bags Stomacher 400 35 cm x 10 cm x 22 cm
142851 Bags Stomacher 400 lab blenders 8 cm x 10.5 cm x 19 cm
142854 Bags Stomacher 3500 15 cm x 42 cm x 35 cm
142852 Bags Stomacher 80Biomaster 30 x 11 x 12 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 142853 Supplier Seward™ Supplier No. 030021126
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Seward has developed a full range of Stomacher™ Accessories to help provide a safe and efficient working environment in your laboratory.

Help to organize sample bags

  • High-grade plastic polymer construction
  • Provide perfect storage for Stomacher bag sample handling for a cleaner, more productive work environment
  • Perfect for loading closure bags pre-enrichment into the incubator
  • Easily dismantled and cleaned with most lab disinfectants (not autoclavable)

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Bags
Material Polymer
Positions 10
Shape Rectangular
Sterility Non-sterile
For Use With (Application) To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment) Stomacher 400
No. per Pack 1
Quantity 1
Dimensions (L x W x H) 35 cm x 10 cm x 22 cm
Product Type Bag Rack
Show More Show Less